missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, Reagent Grade, ≥99%, Solstice

Product Code. 15647940 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 1 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15647940 2.5 L Glass bottle 2.5 L
15667940 500 mL Glass bottle 500mL
15637940 1 L Glass bottle 1 L
15657940 4 x 1 L Glass bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15647940 Supplier Fluka Supplier No. M326312.5L
  • ONLINE EXCLUSIVE PRICE
    153.00 €
    125.13 € / 2.5 L
    Save 27.87 €
    18% Off
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Melting Point -160°C
Color Colorless
Density 0.62g/mL at 25°C
Boiling Point 28°C (1013 hPa)
Flash Point −51°C (−59.8°F)
Ignition Point 788°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)2
Quantity 2.5 L
UN Number UN1265
Beilstein 1730723
Solubility Information Insoluble in water
Vapor Density 2.6 (vs air)
Formula Weight 72.15g/mol
Vapor Pressure 11.17psi (20°C)
Percent Purity ≥99%
Chemical Name or Material 2-Methylbutane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylbutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.