missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, ≥99%, ReagentPlus™, Honeywell™

Product Code. 15673650 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15673650 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15673650 Supplier Honeywell Research Chemicals Supplier No. 3204041L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 78-78-4
Melting Point -160°C
Color Colorless
Density 0.62g/mL at 25°C
Boiling Point 28°C (1013 hPa)
Flash Point −51°C (−59.8°F)
Ignition Point 788°F
Odor Characteristic
Packaging Glass bottle
Molecular Formula C5H12
Linear Formula CH3CH2CH(CH3)2
MDL Number MFCD00009338
Quantity 1 L
UN Number UN1265
Beilstein 1730723
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
Solubility Information Insoluble in water
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
SMILES CCC(C)C
IUPAC Name 2-methylbutane
Molecular Weight (g/mol) 72.15
PubChem CID 6556
Vapor Density 2.6 (vs air)
ChEBI CHEBI:30362
Formula Weight 72.15g/mol
Vapor Pressure 11.17psi (20°C)
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material 2-Methylbutane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.