missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, for HPLC
GREENER_CHOICE

Product Code. 10526494 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10526494 1 L Glass bottle 1 L
10602265 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10526494 Supplier Thermo Scientific Supplier No. 427730010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 78-78-4 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Absorbance 0.01 max. 1 cm cell vs HPLC−grade water at 240nm, 0.01 max. 1 cm cell vs HPLC−grade water at 400nm, 0.07 max. 1 cm cell vs HPLC−grade water at 220nm, 0.3 max. 1 cm cell vs HPLC−grade water at 210nm, 1 max. 1 cm cell vs HPLC grade water at 195nm
Melting Point -160.0°C
Density 0.6200g/mL
Boiling Point 30.0°C
Flash Point −51°C
Packaging Glass bottle
Linear Formula C2H5CH(CH3)2
Refractive Index 1.3530 to 1.3550
Quantity 1 L
Residue after Evaporation 0.0003% max.
Specific Gravity 0.62
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol
Formula Weight 72.15
Percent Purity ≥99.0%
Grade HPLC
Physical Form Liquid
Chemical Name or Material 2-Methylbutane, For HPLC
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.