missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutylamine, 98%

Product Code. 10504531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10504531 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10504531 Supplier Thermo Scientific Acros Supplier No. 205280050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 96-15-1 | C5H13N | 87.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96-15-1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7300g/mL
Boiling Point 94°C to 97°C
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H13N
Linear Formula C2H5CH(CH3)CH2NH2
Refractive Index 1.4106 to 1.4126
MDL Number MFCD00008147
Quantity 5 mL
Beilstein 04, III, 342
Specific Gravity 0.73
Synonym 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
InChI Key VJROPLWGFCORRM-UHFFFAOYSA-N
SMILES CCC(C)CN
IUPAC Name 2-methylbutan-1-amine
Molecular Weight (g/mol) 87.16
PubChem CID 7283
Formula Weight 87.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylbutylamine, 98%
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • 2-Methylbutylamine
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.