missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexylamine, cis + trans, 97%

Product Code. 11387645
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387645 10 g 10g
11397645 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387645 Supplier Thermo Scientific Alfa Aesar Supplier No. L02285.09
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7003-32-9 | C7H15N | 113.20 g/mol

2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7003-32-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001492
InChI Key FEUISMYEFPANSS-UHFFFAOYNA-N
Synonym 2-methylcyclohexylamine, cyclohexanamine, 2-methyl, o-methylcyclohexylamine, cyclohexylamine, 2-methyl, 2-methylcyclohexanamine, 2-methylcyclohexylamine,c&t, hexahydro-o-toluidine, 2methylcyclohexylamine, 2-methyl-cyclohexylamine, 2-methylcyclohexyl amine
PubChem CID 23432
IUPAC Name 2-methylcyclohexan-1-amine
SMILES CC1CCCCC1N

Specifications

Density 0.858
Boiling Point 149°C to 150°C
Flash Point 21°C (69°F)
Refractive Index 1.4565
Quantity 10 g
UN Number UN2733
Beilstein 2689073
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.2
Percent Purity 97%
Chemical Name or Material 2-Methylcyclohexylamine, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.