missing translation for 'onlineSavingsMsg'
Learn More

2-(Methyldithio)isobutyraldehyde, 95%

Product Code. 11915741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915741 5 g Glass bottle 5g
10104880 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915741 Supplier Thermo Scientific Acros Supplier No. 265150050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67952-60-7 | C5H10OS2 | 150.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67952-60-7
Molecular Formula C5H10OS2
Molecular Weight (g/mol) 150.25
InChI Key VLBWEJJOETYCSE-UHFFFAOYSA-N
Synonym 2-methyldithio isobutyraldehyde, 2-methyl-2-methyldithio propanal, unii-3d5g4az6l5, propanal, 2-methyl-2-methyldithio, 2-methyldithio-isobutyraldehyde, alpha-methyldithio isobutyraldehyde, 2-methyl-2-methyldisulfanyl propanal, 2-methyl-2-methyldithio propionaldehyde, 2-methyldisulfanyl isobutyraldehyde, cocoa propanal
PubChem CID 106217
IUPAC Name 2-methyl-2-(methyldisulfanyl)propanal
SMILES CC(C)(C=O)SSC

Specifications

Color Yellow
Density 1.1000g/mL
Boiling Point 46.0°C (1.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5230
Quantity 5 g
Specific Gravity 1.1
Solubility Information Solubility in water: practically insoluble
Formula Weight 150.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(Methyldithio)isobutyraldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Methyldithio)isobutyraldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.