missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran, 99%, stabilized

Product Code. 10046183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10046183 250 mL Glass bottle 250mL
10081140 5 mL Glass bottle 5mL
10552521 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10046183 Supplier Thermo Scientific Acros Supplier No. 126832500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 534-22-5 | C5H6O | 82.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-22-5 , 123-31-9
MDL Number MFCD00003248
InChI Key VQKFNUFAXTZWDK-UHFFFAOYSA-N
Synonym methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
PubChem CID 10797
IUPAC Name 2-methylfuran
SMILES CC1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -88.7°C
Color Yellow
Density 0.9100g/mL
Boiling Point 63.0°C to 66.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H6O
Refractive Index 1.4320 to 1.4340
Quantity 250 mL
Beilstein 17, 36
Fieser 01,682; 14,209
Specific Gravity 0.91
Solubility Information Solubility in water: 0.3g/100mL(20 c). Other solubilities: miscible with most common organic solvents
Stabilizer 0.01% hydroquinone
Molecular Weight (g/mol) 82.1
Viscosity 4 mPa.s (25°C)
Formula Weight 82.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylfuran, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.