missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexane, 99%

Product Code. 10503101 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10503101

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-76-4 | C7H16 | 100.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-76-4
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009487
InChI Key GXDHCNNESPLIKD-UHFFFAOYSA-N
Synonym hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl
PubChem CID 11582
IUPAC Name 2-methylhexane
SMILES CCCCC(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -118.0°C
Color Colorless
Density 0.6700g/mL
Boiling Point 90.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH(CH3)2
Refractive Index 1.3835 to 1.3855
Quantity 5 mL
Beilstein 01, 156
Specific Gravity 0.67
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,miscible with ether,acetone
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylhexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylhexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.