missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexane, 99%

Product Code. 10722685 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10722685 25 mL Glass bottle 25mL
10503101 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.
Código de producto. 10722685 Proveedor Thermo Scientific Acros N.º de proveedor 126910250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.

CAS: 591-76-4 | C7H16 | 100.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 591-76-4
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009487
InChI Key GXDHCNNESPLIKD-UHFFFAOYSA-N
Synonym hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl
PubChem CID 11582
IUPAC Name 2-methylhexane
SMILES CCCCC(C)C

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -118.0°C
Color Colorless
Density 0.6700g/mL
Boiling Point 90.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH(CH3)2
Refractive Index 1.3835 to 1.3855
Quantity 25 mL
Beilstein 01, 156
Specific Gravity 0.67
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,miscible with ether,acetone
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylhexane
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • 2-Methylhexane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.