missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentan-2,4-Diol, Extra Pure, SLR, Fisher Chemical

Product Code. 10538060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10538060 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10538060 Supplier Fisher Chemical Supplier No. M/5050/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 107-41-5
Melting Point -40°C
Boiling Point 197°C
Packaging Amber glass bottle
Molecular Formula C6H14O2
MDL Number 4547
Quantity 500 mL
Synonym hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
InChI Key SVTBMSDMJJWYQN-UHFFFAOYSA-N
SMILES CC(CC(C)(C)O)O
IUPAC Name 2-methylpentane-2,4-diol
Molecular Weight (g/mol) 118.176
PubChem CID 7870
ChEBI CHEBI:62995
Formula Weight 118.18g/mol
Physical Form Viscous Liquid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2,4-pentanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.