missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentane, 99+%

Product Code. 11397925
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397925 100 mL 100mL
11307935 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11397925 Supplier Thermo Scientific Alfa Aesar Supplier No. L03001.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 107-83-5 | C6H14 | 86.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Methylpentane is employed as a raw material, rubber solvent and vegetable oil extraction solvent. It is also used as a solvent in adhesives. Further, it is used as an intermediate in organic synthesis and finds application in food, preservatives, cosmetics, pharmaceuticals, beverages and flavor enhancer.

Solubility
Miscible with alcohol, ether, acetone and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-83-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C

Specifikationer

Melting Point -154°C
Density 0.653
Boiling Point 61°C to 62°C
Flash Point −40°C (−40°F)
Odor Petroleum-like
Refractive Index 1.371
Quantity 100 mL
UN Number UN1208
Beilstein 1730735
Solubility Information Miscible with alcohol,ether,acetone and chloroform. Immiscible with water.
Formula Weight 86.18
Percent Purity ≥99%
Chemical Name or Material 2-Methylpentane
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 2-Methylpentane
Mention d'avertissement
  • Danger
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: 2-Methylpentane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.