missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperidine, 99%

Product Code. 10795391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795391 100 mL Glass bottle 100mL
10502721 5 mL Glass bottle 5mL
10676902 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10795391 Supplier Thermo Scientific Acros Supplier No. 127491000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 109-05-7 | C6H13N | 99.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-05-7
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00005982
InChI Key NNWUEBIEOFQMSS-UHFFFAOYNA-N
Synonym 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline
PubChem CID 7974
IUPAC Name 2-methylpiperidine
SMILES CC1CCCCN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -5°C to 4°C
Color Yellow
Density 0.8440g/mL
Boiling Point 118°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4459 to 1.4479
Quantity 100 mL
Beilstein 20, 95
Specific Gravity 0.844
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with ethanol
Formula Weight 99.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylpiperidine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.