missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpropyl-d9 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methylpropyl alcohol-d9 |
| CAS | 850209-54-0 |
| Molecular Formula | C4 2H9 H O |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C4H10O/c1-4(2)3-5/h4-5 H,3H2,1-2H3/i1D3,2D3,3D2,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-1-ol |
| Molecular Weight (g/mol) | 83.18 |
| Formula Weight | 83.1297 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction