missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrimidine-5-carboxylic acid, 97%

Product Code. 15481549
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15481549

Brand: Thermo Scientific Alfa Aesar H64375.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5194-32-1 | C6H6N2O2 | 138.126 g/mol

It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5194-32-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD08235210
InChI Key NMGIXZFBQPETOK-UHFFFAOYSA-N
Synonym 2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
PubChem CID 13670875
IUPAC Name 2-methylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C=N1)C(=O)O

Specifications

Melting Point 203°C to 205°C
Quantity 1 g
Beilstein 508773
Solubility Information Slightly soluble in water.
Formula Weight 138.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.