missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™

Product Code. 15522333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15522333 1 g 1g
15532333 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15522333 Supplier Thermo Scientific Acros Supplier No. 458070010
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 54677-53-1
Molecular Formula C5H11N·HCl
Molecular Weight (g/mol) 121.61
InChI Key JNEIFWYJFOEKIM-UHFFFAOYSA-N
Synonym 2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
PubChem CID 21907341
IUPAC Name 2-methylpyrrolidine;hydrochloride
SMILES CC1CCCN1.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 121.61
Percent Purity 97%
Chemical Name or Material 2-Methylpyrrolidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.