missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoline, TRC

Product Code. 30137019
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137019 500 mg 500mg
17902945 1 g 1g
17992545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137019 Supplier Toronto Research Chemicals Supplier No. TRCM204951500MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-63-4
Molecular Formula C10 H9 N
Molecular Weight (g/mol) 143.19
Synonym Quinaldine (8CI), 2-Methylquinoline, Khinaldin, NSC 3397
IUPAC Name 2-methylquinoline
SMILES Cc1ccc2ccccc2n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H9N/c1-8-6-7-9-4-2-3-5-10(9)11-8/h2-7H,1H3
Formula Weight 143.0735
Percent Purity >95%
Chemical Name or Material 2-Methylchinoline
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.