missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoxaline 1,4-Dioxide, TRC

Product Code. 30478234
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478234 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478234 Supplier Toronto Research Chemicals Supplier No. TRCM326390100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylquinoxaline 1,4-Dioxide
CAS 6639-86-7
Molecular Formula C9H8N2O2
Quantity 100 mg
Synonym 2-Methylquinoxaline N,N'-dioxide, 2-Methylquinoxaline di-N-oxide, NSC 47509, NSC 47510, NSC 48949
InChI Formula InChI=1S/C9H8N2O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3
SMILES Cc1c[n+]([O-])c2ccccc2[n+]1[O-]
IUPAC Name 2-methyl-1,4-dioxidoquinoxaline-1,4-diium
Molecular Weight (g/mol) 176.17
Formula Weight 176.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.