missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfinyl)benzeneboronic acid, 97%

Product Code. 15459967
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459967 250 mg 250mg
15449967 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459967 Supplier Thermo Scientific Alfa Aesar Supplier No. H53209.MD
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 850567-97-4 | C7H9BO3S | 184.02 g/mol

It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-97-4
Molecular Formula C7H9BO3S
Molecular Weight (g/mol) 184.02
MDL Number MFCD03095261
InChI Key PHORKVSBWZGTEX-UHFFFAOYNA-N
Synonym 2-methylsulfinyl phenylboronic acid, 2-methylsulfinyl phenyl boronic acid, 2-methylsulfinylphenyl boronic acid, 2-methylsulphinyl benzeneboronic acid, 2-methanesulfinylphenylboronic acid, 1-borono-2-methylsulphinyl benzene, 2-methylsulfinyl benzeneboronic acid, pubchem1874, acmc-209q0z
PubChem CID 2773529
IUPAC Name (2-methylsulfinylphenyl)boronic acid
SMILES CS(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 118°C to 122°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 184.02
Percent Purity 97%
Chemical Name or Material 2-(Methylsulfinyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.