missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl) Phenothiazine, TRC

Product Code. 30137837
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137837 100 mg 100mg
30137109 1 g 1g
30009109 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137837 Supplier Toronto Research Chemicals Supplier No. TRCM326625100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23503-68-6
Molecular Formula C13 H11 N O2 S2
Molecular Weight (g/mol) 277.36
Synonym 10H-Phenothiazine, 2-(methylsulfonyl)-, Phenothiazine, 2-(methylsulfonyl)- (6CI,8CI)
IUPAC Name 2-methylsulfonyl-10H-phenothiazine
SMILES CS(=O)(=O)c1ccc2Sc3ccccc3Nc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H11NO2S2/c1-18(15,16)9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)17-13/h2-8,14H,1H3
Formula Weight 277.0231
Chemical Name or Material 2-Methylsulfonyl-10H-phenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.