missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)-1,3-benzothiazol-6-amine, 97%, Thermo Scientific™

Product Code. 10792493
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792493 1 g Amber Glass Bottle 1g
10101082 250 mg Amber Glass Bottle 250mg
10347114 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10792493 Supplier Thermo Scientific Maybridge Supplier No. KM06901DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25706-29-0
Molecular Formula C8H8N2S2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00086478
InChI Key GYCJZSAIEMSFAV-UHFFFAOYSA-N
PubChem CID 274151
IUPAC Name 2-methylsulfanyl-1,3-benzothiazol-6-amine
SMILES CSC1=NC2=CC=C(N)C=C2S1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-methylthio-1,3-benzothiazol-6-amine, 2-methylsulfanyl-1,3-benzothiazol-6-amine, 2-methylthio benzo d thiazol-6-amine, 6-benzothiazolamine, 2-methylthio, 2-methylsulfanyl-benzothiazol-6-ylamine, 6-benzothiazolamine,2-methylthio, 6-amino-2-methylthio benzothiazole, 2-methylthiobenzothiazole-6-ylamine, enamine_005325, 6-amino-2-methylthio-1,3-benzothiazole
Formula Weight 196.3
Percent Purity 97%
Chemical Name or Material 2-(Methylthio)-1, 3-benzothiazol-6-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.