missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethylamine, 95%

Product Code. 11470008
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470008 5 g 5g
11460008 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470008 Supplier Thermo Scientific Alfa Aesar Supplier No. B22941.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18542-42-2 | C3H9NS | 91.172 g/mol

2-(Methylthio)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethylamine is a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18542-42-2
Molecular Formula C3H9NS
Molecular Weight (g/mol) 91.172
MDL Number MFCD00014825
InChI Key CYWGSFFHHMQKET-UHFFFAOYSA-N
Synonym 2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
PubChem CID 87697
IUPAC Name 2-methylsulfanylethanamine
SMILES CSCCN

Specifications

Density 0.974
Boiling Point 146°C to 148°C
Flash Point 36°C (96°F)
Odor Amine-like
Linear Formula CH3S(CH2)2NH2
Refractive Index 1.4965
Quantity 5 g
UN Number UN2734
Beilstein 1731099
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 91.18
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.