missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)thiophene, 97%

Product Code. 11469323
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469323 5 g 5g
11459323 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469323 Supplier Thermo Scientific Alfa Aesar Supplier No. A11763.06
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 5780-36-9 | C5H6S2 | 130.223 g/mol

2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5780-36-9
Molecular Formula C5H6S2
Molecular Weight (g/mol) 130.223
MDL Number MFCD00052382
InChI Key ZLSMPEVZXWDWEK-UHFFFAOYSA-N
PubChem CID 79844
IUPAC Name 2-methylsulfanylthiophene
SMILES CSC1=CC=CS1

Specifications

Boiling Point 82°C to 84°C (15 mmHg)
Odor Stench
Refractive Index 1.596
Quantity 5 g
UN Number UN3334
Beilstein 106272
Synonym 2-methylthio thiophene, thiophene, 2-methylthio, 2-methylsulfanyl thiophene, 2-methylthiothiophene, 2-mehtylthio thiophene, thiophene,2-methylthio, acmc-20aomi, 2-methylthio-thiophene, 2-methylmercaptothiophene, 2-methylsulfanyl thiophene #
Formula Weight 130.23
Percent Purity 97%
Chemical Name or Material 2- (Methylthio)thiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.