missing translation for 'onlineSavingsMsg'
Learn More

2-Methylvaleric acid, 98+%

Product Code. 11491548
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491548 100 g 100g
11401558 500 g 500g
11411558 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491548 Supplier Thermo Scientific Alfa Aesar Supplier No. A13873.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-61-0 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
(R,S)-2-methylvaleric acid is an intermediate in the synthesis of branched-chain. 2-Methylpentanoic acid (2-Methylvaleric acid) was used as an internal standard for gas chromatographic analysis of microbial end products.

Solubility
Soluble in water(13g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-61-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002671
InChI Key OVBFMEVBMNZIBR-UHFFFAOYSA-N
Synonym 2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754
PubChem CID 7341
IUPAC Name 2-methylpentanoic acid
SMILES CCCC(C)C(=O)O

Specifications

Melting Point -85°C
Density 0.93
Boiling Point 196°C to 197°C
Flash Point 91°C (195°F)
Odor Unpleasant
Refractive Index 1.414
Quantity 100 g
UN Number UN3265
Beilstein 1720655
Solubility Information Soluble in water(13g/L).
Formula Weight 116.16
Percent Purity ≥98%
Chemical Name or Material 2-Methylvaleric acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylvaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methylvaleric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.