missing translation for 'onlineSavingsMsg'
Learn More

2-Methylvaleryl chloride, 98%

Product Code. 11357805
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357805 25 g 25g
11347805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357805 Supplier Thermo Scientific Alfa Aesar Supplier No. L02725.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5238-27-7 | C14H12N2O3S | 288.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5238-27-7
Molecular Formula C14H12N2O3S
Molecular Weight (g/mol) 288.32
MDL Number MFCD00013656
InChI Key NADADZQYJBFZGI-FLIBITNWSA-N
Synonym 2-methylvaleryl chloride, 2-methylvaleroyl chloride, 2-methylvalerylchloride, .alpha.-methyl valeroyl chloride, acmc-20anpk, 2-methyvaleryl chloride, pubchem21794, 2-methyl valeryl chloride, 2-methyl-valeryl chloride, 2-methyl pentanoyl chloride
PubChem CID 107385
SMILES CC1=CC=C(C=C1)C(\O)=C\C(=O)C(=O)NC1=NC=CS1

Specifications

Density 0.978
Boiling Point 140°C to 144°C
Flash Point 37°C (98°F)
Odor Acrid
Refractive Index 1.4239
Quantity 25 g
UN Number UN2920
Beilstein 1743993
Sensitivity Moisture sensitive
IUPAC Name 2-methylpentanoyl chloride
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material 2-Methylvaleryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.