missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10154104
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10154104 250 mg Amber Glass Bottle 250mg
10131174 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10154104 Supplier Thermo Scientific Maybridge Supplier No. CC62801CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 906353-04-6
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.24
MDL Number MFCD09702410
InChI Key JXQGQYNBVNKCAK-UHFFFAOYSA-N
PubChem CID 24229698
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1=CSC(=N1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid, 2-morpholino-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-morpholinyl, 2-morpholinothiazole-4-carboxylic acid, 4-4-carboxy-1,3-thiazol-2-yl morpholine, 4-thiazolecarboxylicacid,2-4-morpholinyl, 4-carboxy-2-morpholin-4-yl-1,3-thiazole, 4-thiazolecarboxylic acid, 2-4-morpholinyl, 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 2-Morpholino-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.