missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoaniline, 98%, BLD Pharm™

Product Code. 31103543
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31103543 10 g
31098134 5 g
31092812 100 g
31092952 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31103543 Supplier BLD Pharm Supplier No. BD1221310g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5585-33-1
Molecular Formula C10H14N2O
MDL Number MFCD00047408
Quantity 10 g
SMILES NC1=CC=CC=C1N2CCOCC2
Molecular Weight (g/mol) 178.23
Percent Purity 98%
Chemical Name or Material 2-Morpholinoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.