missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10321925
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10321925 5 g 5g
10019471 1 g 1g
10374912 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10321925 Supplier Thermo Scientific Maybridge Supplier No. SEW05383DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58028-76-5
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00662562
InChI Key GTTAEWVBVHSDLX-UHFFFAOYSA-N
Synonym 2-morpholinobenzaldehyde, 2-morpholin-4-yl-benzaldehyde, 2-4-morpholino benzaldehyde, 2-morpholin-4-yl benzaldehyde, benzaldehyde, 2-4-morpholinyl, morpholinobenzaldehyde, pubchem16276, acmc-1b05v, 2-morpholinobenzenecarbaldehyde, 4-2-formylphenyl morpholine
PubChem CID 2737010
IUPAC Name 2-morpholin-4-ylbenzaldehyde
SMILES O=CC1=CC=CC=C1N1CCOCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 2-Morpholinobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.