missing translation for 'onlineSavingsMsg'
Learn More

2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%

Product Code. 10672803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10672803 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10672803 Supplier Thermo Scientific Acros Supplier No. 360485000
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 182287-51-0 | C13H23NO5 | 273.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 182287-51-0
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C13H23NO5
MDL Number MFCD02683139
Quantity 500 mg
Synonym 2-n-boc-amino-3-4-tetrahydropyranyl propionic acid, 2-tert-butoxycarbonyl amino-3-tetrahydro-2h-pyran-4-yl propanoic acid, 2-boc-3-tetrahydropyran-4-yl-dl-alanine, n-tert-butoxycarbonyl-3-oxan-4-ylalanine, 2-tert-butoxycarbonylamino-3-tetrahydro-pyran-4-yl-propionic acid, 2-tert-butoxy carbonyl amino-3-oxan-4-yl propanoic acid, 2-tert-butoxycarbonyl amino-3-oxan-4-yl propanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-oxan-4-yl propanoic acid, alpha-boc-amino tetrahydropyran-4-propanoic acid
InChI Key NKYZORHKIYSSEL-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC(CC1CCOCC1)C(=O)O
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(oxan-4-yl)propanoic acid
Molecular Weight (g/mol) 273.33
PubChem CID 2734406
Formula Weight 273.33
Percent Purity 95%
Physical Form Chunks
Chemical Name or Material 2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-N-BOC-Amino-3-(4-tetrahydropyranyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.