missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Butoxycarbonylpiperidine)-5-methyl-1,3-thiazol-4-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10533403 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533403 1 g Amber Glass Bottle 1g
10492803 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533403 Supplier Thermo Scientific Supplier No. MO07259DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-88-3
Molecular Formula C16H24N2O4S
Molecular Weight (g/mol) 340.438
InChI Key IHHZQULPYDMAOP-UHFFFAOYSA-N
Synonym 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-thiazol-4-yl acetic acid, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl aceticacid, 2-5-methyl-2-n-tert-butoxycarbonylpiperidine-1,3-thiazol-4-yl acetic acid, 2-5-methyl-2-piperidine-n-boc protected-1,3-thiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-t, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-y, 2-1-butoxycarbonylpiperidine-5-methyl-1,3-thiazol-4-yl acetic acid
PubChem CID 2794810
IUPAC Name 2-[5-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazol-4-yl]acetic acid
SMILES CC1=C(N=C(S1)C2CCN(CC2)C(=O)OC(C)(C)C)CC(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 340.44
Percent Purity 97%
Chemical Name or Material {2-[1-(tert-Butoxycarbonyl)piperidin-4-yl]-5-methyl-1, 3-thiazol-4-yl}acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.