missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexylpyridine, 98%

Product Code. 11455113
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455113 25 g 25g
11445113 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455113 Supplier Thermo Scientific Alfa Aesar Supplier No. A12772.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1129-69-7 | C11H17N | 163.264 g/mol

2-n-Hexylpyridine is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Hexylpyridine is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
This product is relativelyunstable under normal temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-69-7
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00023552
InChI Key NZLJDTKLZIMONR-UHFFFAOYSA-N
Synonym pyridine, 2-hexyl, 2-n-hexylpyridine, 2-n-hexyl pyridine, acmc-20antt, pyridine, 2-n-hexyl, 5-20-06-00161 beilstein handbook reference
PubChem CID 70797
IUPAC Name 2-hexylpyridine
SMILES CCCCCCC1=CC=CC=N1

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 163.27
Percent Purity 98%
Chemical Name or Material 2-n-Hexylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.