missing translation for 'onlineSavingsMsg'
Learn More

2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine, TRC

Product Code. 30331995
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331995 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331995 Supplier Toronto Research Chemicals Supplier No. TRCB58556510G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine
CAS 145100-51-2
Molecular Formula C7H3ClF6N2O4S2
Quantity 10 g
Synonym N-(5-Chloro-2-pyridinyl)-1,1,1-trifluoro-N-[(trifluoromethyl)sulfonyl]methanesulfonamide, Comins' Reagent, N,N-Bis(trifluoromethylsulfonyl)-5-chloro-2-pyridinamine, N,N-Bis(trifluoromethylsulfonyl)-5-chloro-2-pyridylamine, N-(5-Chloro-2-pyridyl)triflimide, N-(5-Chloropyridin-2-yl)-1,1,1-trifluoro-N-[(trifluoromethyl)sulfonyl]methanesulfonamide, NSC 683533
InChI Formula InChI=1S/C7H3ClF6N2O4S2/c8-4-1-2-5(15-3-4)16(21(17,18)6(9,10)11)22(19,20)7(12,13)14/h1-3H
SMILES FC(F)(F)S(=O)(=O)N(c1ccc(Cl)cn1)S(=O)(=O)C(F)(F)F
IUPAC Name N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
Molecular Weight (g/mol) 392.68
Formula Weight 391.9127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.