missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Propylthio)nicotinic acid, 97%, Thermo Scientific™

Product Code. 11427576
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427576 100 g 100g
11407576 5 g 5g
11417576 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427576 Supplier Thermo Scientific Alfa Aesar Supplier No. A14623.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175135-22-5
Molecular Formula C9H10NO2S
Molecular Weight (g/mol) 196.24
MDL Number MFCD00051682
InChI Key CSMDLVRDBYKTAH-UHFFFAOYSA-M
Synonym 2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
PubChem CID 2774526
IUPAC Name 2-propylsulfanylpyridine-3-carboxylic acid
SMILES CCCSC1=NC=CC=C1C([O-])=O

Specifications

Melting Point 159°C to 163°C
Quantity 100 g
Formula Weight 197.26
Percent Purity 97%
Chemical Name or Material 2-(n-Propylthio)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.