missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalen-2-yl-piperazine, Thermo Scientific™

Product Code. 10812231
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10812231 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10812231 Supplier Thermo Scientific Maybridge Supplier No. MAY00168DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 904816-32-6
Packaging Amber Glass Bottle
Molecular Formula C14H16N2
MDL Number MFCD05864679
Quantity 1 g
Synonym 2-naphthalen-2-yl-piperazine, 2-naphthalen-2-yl piperazine, 2-2-naphthyl piperazine, 2-naphth-2-yl piperazine, acmc-20a0jo, 2-phthalen-2-yl-piperazine
InChI Key DNMNNHMPDTUAFG-UHFFFAOYNA-N
SMILES C1CNC(CN1)C1=CC=C2C=CC=CC2=C1
IUPAC Name 2-naphthalen-2-ylpiperazine
Molecular Weight (g/mol) 212.30
PubChem CID 2771839
Formula Weight 212.29
Percent Purity 95%
Chemical Name or Material 2-naphthalen-2-yl-piperazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.