missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoic Acid, TRC

Product Code. 30469586
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469586 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469586 Supplier Toronto Research Chemicals Supplier No. TRCN34564025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Naphthoic Acid
CAS 93-09-4
Molecular Formula C11H8O2
Quantity 25 g
Synonym 2-Naphthalenecarboxylic Acid, β-Naphthoic Acid, 2-Naphthoic Acid (8CI), 2-Carboxynaphthalene, 2-Maythic Acid, 2-Naphthylcarboxylic Acid, Isonaphthoic Acid, NSC 59901, GI 149398X, BRL-19764HT, SKF-58865, Salmeterol related
InChI Formula InChI=1S/C11H8O2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,(H,12,13)
SMILES OC(=O)c1ccc2ccccc2c1
IUPAC Name naphthalene-2-carboxylic acid
Molecular Weight (g/mol) 172.18
Formula Weight 172.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.