missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoyl chloride, 98%

Product Code. 11457946
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457946 10 g 10g
11467946 50 g 50g
11477946 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457946 Supplier Thermo Scientific Alfa Aesar Supplier No. A14046.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2243-83-6 | C11H7ClO | 190.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-83-6
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.626
MDL Number MFCD00004093
InChI Key XNLBCXGRQWUJLU-UHFFFAOYSA-N
Synonym 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
PubChem CID 75246
IUPAC Name naphthalene-2-carbonyl chloride
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)Cl

Specifications

Melting Point 48°C to 52°C
Boiling Point 159°C to 160°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 10 g
UN Number UN3261
Beilstein 907776
Sensitivity Moisture sensitive
Formula Weight 190.63
Percent Purity 98%
Chemical Name or Material 2-Naphthoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.