missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetic acid, 99%

Product Code. 11498760
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498760 5 g 5g
11475633 10 g 10g
11485633 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498760 Supplier Thermo Scientific Alfa Aesar Supplier No. A13528.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 581-96-4 | C12H10O2 | 186.21 g/mol

2-Naphthylacetic acid is a synthetic plant hormone in the auxin family and is an ingredient in many commercial plant rooting horticultural products, it is a rooting agent and used for the vegetative propagation of plants from stem and leaf cutting. It is also used for plant tissue culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthylacetic acid is a synthetic plant hormone in the auxin family and is an ingredient in many commercial plant rooting horticultural products, it is a rooting agent and used for the vegetative propagation of plants from stem and leaf cutting. It is also used for plant tissue culture.

Solubility
Soluble in acetone (50 mg/ml), and methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-96-4
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004126
InChI Key VIBOGIYPPWLDTI-UHFFFAOYSA-N
Synonym 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
PubChem CID 11393
ChEBI CHEBI:37837
IUPAC Name 2-naphthalen-2-ylacetic acid
SMILES OC(=O)CC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Beilstein 973396
Solubility Information Soluble in acetone (50mg/ml),and methanol. Insoluble in water.
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Naphthylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.