missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-p-phenylenediamine 95-99%, ACROS Organics™

Product Code. 10499240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10499240 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10499240 Supplier Thermo Scientific Acros Supplier No. 128801000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5307-14-2
CAS Min % 95.0
CAS Max % 100.0
Melting Point 134.0°C to 140.0°C
Color Brown to Black
Infrared Spectrum Authentic
Assay Percent Range 95-99%
Packaging Glass bottle
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
MDL Number MFCD00007903
Quantity 100 g
Beilstein 13, 120
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hcl
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N
IUPAC Name 2-nitrobenzene-1,4-diamine
Molecular Weight (g/mol) 153.141
PubChem CID 4338370
ChEBI CHEBI:76394
Formula Weight 153.14
Percent Purity 95 to 99%
Physical Form Powder
Chemical Name or Material 2-Nitro-p-phenylenediamine, 95-99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitro-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.