missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-p-phenylenediamine, 95%

Product Code. 11455477
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455477 500 g 500g
11435477 25 g 25g
11445477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455477 Supplier Thermo Scientific Alfa Aesar Supplier No. B23477.36
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5307-14-2 | C6H7N3O2 | 153.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5307-14-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007903
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
PubChem CID 4338370
ChEBI CHEBI:76394
IUPAC Name 2-nitrobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N

Specifications

Melting Point 134°C to 137°C
Quantity 500 g
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 2-Nitro-p-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.