missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroaniline, ≥99.0% (HPLC), Honeywell™ Fluka™

Product Code. 15666900 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15666900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15666900 Supplier Honeywell-Fluka Supplier No. 72660100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator (in nonaqueous solvents),≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 70°C to 72°C
Boiling Point 284°C
CAS 88-74-4
Molecular Formula C6H6N2O2
Linear Formula O2NC6H4NH2
MDL Number MFCD00007687
UN Number UN1661
Beilstein 509275
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]
IUPAC Name 2-nitroaniline
Molecular Weight (g/mol) 138.126
PubChem CID 6946
Formula Weight 138.12g/mol
Percent Purity ≥99.0% (HPLC)
Packaging Glass bottle
Quantity 100 g
Flash Point 167°C (332.6°F)
Physical Form Crystalline
Chemical Name or Material 2-Nitroaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.