missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonyl chloride, 97%

Product Code. 11457084
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457084 100 g 100g
11447084 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457084 Supplier Thermo Scientific Alfa Aesar Supplier No. B21522.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1694-92-4 | C6H4ClNO4S | 221.611 g/mol

2-Nitrobenzenesulfonyl chloride acts as a reagent to form 2-nitrobenzenesulfonyl derivatives of alcohols and amines. It is also involved in the selective cleavage with thiophenol without displacement of the nitro group. Further, it is used in the preparation of renin inhibitors. In addition to this, it finds application as an anticonvulsant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzenesulfonyl chloride acts as a reagent to form 2-nitrobenzenesulfonyl derivatives of alcohols and amines. It is also involved in the selective cleavage with thiophenol without displacement of the nitro group. Further, it is used in the preparation of renin inhibitors. In addition to this, it finds application as an anticonvulsant.

Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride, ethyl acetate and dimethylformamide. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1694-92-4
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007430
InChI Key WPHUUIODWRNJLO-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
PubChem CID 15540
IUPAC Name 2-nitrobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl

Specifications

Melting Point 64°C to 69°C
Assay Percent Range 0.97
Quantity 100 g
UN Number UN3261
Beilstein 521155
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene,tetrahydrofuran,methylene chloride,ethyl acetate and dimethylformamide. Insoluble in water.
Formula Weight 221.62
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.