missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzhydrazide, 98%

Product Code. 11479247
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479247 25 g 25g
11469247 5 g 5g
11489247 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479247 Supplier Thermo Scientific Alfa Aesar Supplier No. B21330.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 606-26-8 | C7H7N3O3 | 181.151 g/mol

The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

Solubility
Soluble in methanol(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-26-8
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007598
InChI Key LYGGDXLOJMNFBV-UHFFFAOYSA-N
Synonym 2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
PubChem CID 3781218
IUPAC Name 2-nitrobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point 119°C to 123°C
Quantity 25 g
Beilstein 394754
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.