missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11925611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11925611 10 g Glass bottle 10g
11945611 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925611 Supplier Thermo Scientific Acros Supplier No. 128470100
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-24-8
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007044
InChI Key SWBDKCMOLSUXRH-UHFFFAOYSA-N
Synonym o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
PubChem CID 11922
IUPAC Name 2-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110.8°C to 111.8°C
Color Yellow
Boiling Point 165.0°C (16.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 10 g
Beilstein 09,374
Solubility Information Solubility in water: insoluble
Formula Weight 148.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.