missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzotrifluoride, TRC

Product Code. 30468193
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468193 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468193 Supplier Toronto Research Chemicals Supplier No. TRCN49501850MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitrobenzotrifluoride
CAS 384-22-5
Molecular Formula C7 H4 F3 N O2
Quantity 50 mg
Synonym Toluene','α,α,α-trifluoro-o-nitro- (6CI,7CI,8CI), 1-Nitro-2-(trifluoromethyl)benzene, 2-(Trifluoromethyl)nitrobenzene, 2-Nitro-α,α,α-trifluorotoluene, 2-Nitrobenzotrifluoride, o-(Trifluoromethyl)nitrobenzene, o-Nitrobenzotrifluoride, α,α,α-Trifluoro-o-nitrotoluene, Benzene','1-nitro-2-(trifluoromethyl)-
InChI Formula InChI=1S/C7H4F3NO2/c8-7(9,10)5-3-1-2-4-6(5)11(12)13/h1-4H
SMILES [O-][N+](=O)c1ccccc1C(F)(F)F
IUPAC Name 1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 191.107
Formula Weight 191.0194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.