missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, 97%

Product Code. 10102374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10102374 5 g Glass bottle 5g
10684925 25 g Glass bottle 25g
10192944 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102374 Supplier Thermo Scientific Acros Supplier No. 397960050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 625-48-9 | C2H5NO3 | 91.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-48-9
Molecular Formula C2H5NO3
Molecular Weight (g/mol) 91.07
MDL Number MFCD00007405
InChI Key KIPMDPDAFINLIV-UHFFFAOYSA-N
Synonym ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
PubChem CID 12252
IUPAC Name 2-nitroethanol
SMILES C(CO)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Yellow
Density 1.2920g/mL
Boiling Point 203.0°C to 206.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NCH2CH2OH
Refractive Index 1.4445
Quantity 5 g
Specific Gravity 1.292
Solubility Information Solubility in water: miscible
Formula Weight 91.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Nitroethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitroethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.