missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, tech. 80%

Product Code. 11413268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413268 5 g 5g
11423268 25 g 25g
11433268 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413268 Supplier Thermo Scientific Alfa Aesar Supplier No. A17681.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 625-48-9 | C2H5NO3 | 91.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-48-9
Molecular Formula C2H5NO3
Molecular Weight (g/mol) 91.066
MDL Number MFCD00007405
InChI Key KIPMDPDAFINLIV-UHFFFAOYSA-N
Synonym ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
PubChem CID 12252
IUPAC Name 2-nitroethanol
SMILES C(CO)[N+](=O)[O-]

Specifications

Melting Point -80°C
Density 1.27
Boiling Point 194°C
Flash Point >110°C (230°F)
Refractive Index 1.4451
Quantity 5 g
Beilstein 1633753
Formula Weight 91.07
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-Nitroethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.