missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroimidazole, 98%

Product Code. 11353137
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353137 100 mg 100mg
11343137 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353137 Supplier Thermo Scientific Alfa Aesar Supplier No. L13546.MC
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol

2-Nitroimidazole, is used as an antibiotic in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitroimidazole, is used as an antibiotic in pharma industry.

Solubility
It is slightly soluble in water. Soluble in acetone, butyl acetate, ethanol, ethyl acetate and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids, acid chlorides, anhydrous acids.. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-73-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005185
InChI Key YZEUHQHUFTYLPH-UHFFFAOYSA-N
Synonym 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
PubChem CID 10701
ChEBI CHEBI:67135
IUPAC Name 2-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=NC=CN1

Specifications

Melting Point ∽287°C (decomposition)
Quantity 100 mg
UN Number UN2811
Beilstein 116444
Merck Index 14,921
Solubility Information It is slightly soluble in water. Soluble in acetone,butyl acetate,ethanol,ethyl acetate and methanol.
Formula Weight 113.08
Percent Purity 98%
Chemical Name or Material 2-Nitroimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.