missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Nitrophenol |
| CAS | 88-75-5 |
| Molecular Formula | C6 H5 N O3 |
| Quantity | 5 g |
| Synonym | Phenol','2-nitro-, Phenol','o-nitro- (8CI), 2-Nitrophenol, 2-Hydroxynitrobenzene, NSC 1552, o-Hydroxynitrobenzene, o-Nitrophenol |
| InChI Formula | InChI=1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H |
| SMILES | Oc1ccccc1[N+](=O)[O-] |
| IUPAC Name | 2-nitrophenol |
| Molecular Weight (g/mol) | 139.11 |
| Formula Weight | 139.0269 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction