missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol, TRC

Product Code. 30468557
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30468557

Brand: Toronto Research Chemicals TRCN496905100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitrophenol
CAS 88-75-5
Molecular Formula C6 H5 N O3
Quantity 100 g
Synonym Phenol','2-nitro-, Phenol','o-nitro- (8CI), 2-Nitrophenol, 2-Hydroxynitrobenzene, NSC 1552, o-Hydroxynitrobenzene, o-Nitrophenol
InChI Formula InChI=1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H
SMILES Oc1ccccc1[N+](=O)[O-]
IUPAC Name 2-nitrophenol
Molecular Weight (g/mol) 139.11
Formula Weight 139.0269
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.