missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl isocyanate, 99%

Product Code. 10522471 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522471 25 g Glass bottle 25g
10319310 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10522471 Supplier Thermo Scientific Acros Supplier No. 296690250
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3320-86-3 | C7H4N2O3 | 164.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3320-86-3
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
InChI Key JRVZITODZAQRQM-UHFFFAOYSA-N
Synonym 2-nitrophenyl isocyanate, o-nitrophenyl isocyanate, benzene, 1-isocyanato-2-nitro, isocyanic acid, o-nitrophenyl ester, 2-nitrophenylisocyanate, 2-nitrobenzenisocyanate, nitrophenyl isocyanate, 2-nitrophenylisocynate, 2-nitro phenyl isocyanate, acmc-1cp5k
PubChem CID 76834
IUPAC Name 1-isocyanato-2-nitrobenzene
SMILES C1=CC=C(C(=C1)N=C=O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 41.0°C
Color Yellow
Boiling Point 135.0°C to 137.0°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NCO
Quantity 25 g
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 164.12
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Nitrophenyl isocyanate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrophenyl isocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.