missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl isothiocyanate, 97%

Product Code. 15448909
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448909 Supplier Thermo Scientific Alfa Aesar Supplier No. L09669.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2719-30-4 | C7H4N2O2S | 180.181 g/mol

2-Nitrophenyl isothiocyanate can react with pyridin-2-ylamine to get 2-nitrophenyl(2-pyridyl)thiourea. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenyl isothiocyanate can react with pyridin-2-ylamine to get 2-nitrophenyl(2-pyridyl)thiourea. It is also used as pharmaceutical intermediates.

Solubility
Hydrolyzes with water.

Notes
Store
TRUSTED_SUSTAINABILITY

Specifications

CAS 2719-30-4
Melting Point 73°C to 76°C
Boiling Point 135°C to 137°C (17 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Molecular Formula C7H4N2O2S
MDL Number MFCD00007092
Quantity 5 g
UN Number UN2923
Beilstein 2719657
Sensitivity Moisture sensitive
Synonym 2-nitrophenyl isothiocyanate, benzene, isothiocyanatonitro, 2-nitrobenzenisothiocyanate, 2-nitrophenylisothiocyanate, o-nitrophenyl isothiocyanate, 1-isothiocyanato-2-nitro-benzene, #
Solubility Information Hydrolyzes with water.
InChI Key CBWJHIXSVFDERH-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N=C=S)[N+](=O)[O-]
IUPAC Name 1-isothiocyanato-2-nitrobenzene
Molecular Weight (g/mol) 180.181
PubChem CID 137690
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 2-Nitrophenyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Nitrophenyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.